Assay Details
| Gene Symbol: Otub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106332.1 | NP_001099802.1 | 2 - 3 | 150 | 69 | |
| GenBank mRNA | BC166771.1 | - | 2 - 3 | 133 | 7934088 | 69 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Otub1
Genomic Map