Assay Details
| Gene Symbol: Cabp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108926.1 | NP_001102396.1 | 1 - 2 | 418 | 70 | |
| XM_006230867.2 | XP_006230929.1 | 1 - 2 | 1723 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cabp4
Genomic Map