Assay Details
| Gene Symbol: Cabp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108926.1 | NP_001102396.1 | 3 - 4 | 586 | 107 | |
| XM_006230867.2 | XP_006230929.1 | 3 - 4 | 1891 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cabp4
Genomic Map