Assay Details
| Gene Symbol: Acta2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031004.2 | NP_112266.1 | 2 - 3 | 183 | 71 | |
| GenBank mRNA | BC158550.1 | - | 2 - 3 | 177 | 9033322 | 71 |
| DN934985.1 | - | 2 - 3 | 212 | 71 | ||
| X06801.1 | - | 2 - 3 | 168 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acta2
Genomic Map