Assay Details
| Gene Symbol: Frmd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008348.1 | NP_001008349.1 | 8 - 9 | 1089 | 67 | |
| XM_006230804.2 | XP_006230866.1 | 8 - 9 | 1113 | 67 | ||
| GenBank mRNA | BC085896.1 | - | 8 - 9 | 1089 | 7193163 | 67 |
| FQ211651.1 | - | 8 - 9 | 1128 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Frmd8
Genomic Map