Assay Details
| Gene Symbol: Cdk2ap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109498.1 | NP_001102968.1 | 1 - 2 | 162 | 133 | |
| XM_006230902.2 | XP_006230964.1 | 1 - 2 | 552 | 133 | ||
| GenBank mRNA | FQ234216.1 | - | 1 - 2 | 164 | 133 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109498.1,XM_006230902.2 transcripts.
Set Membership: |
|

Cdk2ap2
Genomic Map