Assay Details
| Gene Symbol: NEWGENE_1565505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760350.1 | XP_008758572.1 | 1 - 2 | 646 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008760350.1 transcripts.
Set Membership: |
|

NEWGENE_1565505
Genomic Map