Assay Details
| Gene Symbol: NEWGENE_1565505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760350.1 | XP_008758572.1 | 2 - 3 | 1140 | 58 | |
| XM_008760351.1 | XP_008758573.1 | 2 - 3 | 1509 | 58 | ||
More Information
Set Membership: |
|

NEWGENE_1565505
Genomic Map