Assay Details
| Gene Symbol: Psbpc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207593.2 | NP_997476.2 | 2 - 3 | 287 | 147 | |
| GenBank mRNA | BC099748.1 | - | 2 - 3 | 281 | 147 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psbpc2
Genomic Map