Assay Details
| Gene Symbol: Lyc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128494.1 | NP_001121966.1 | 1 - 2 | 385 | 104 | |
| XM_006241392.2 | XP_006241454.1 | 1 - 2 | 209 | 104 | ||
| GenBank mRNA | BC166737.1 | - | 1 - 2 | 385 | 7937244 | 104 |
More Information
Updated Mapping Information
Set Membership: |
|

Lyc2
Genomic Map