Assay Details
| Gene Symbol: Il22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191988.1 | NP_001178917.1 | 1 - 2 | 188 | 66 | |
| GenBank mRNA | DQ630444.1 | - | 1 - 2 | 167 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il22
Genomic Map