Assay Details
| Gene Symbol: LOC100909505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750325.3 | XP_003750373.1 | 349 | 113 | ||
| Gene Symbol: Gns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011989.1 | NP_001011989.1 | 2 - 3 | 252 | 113 | |
| XM_006241396.2 | XP_006241458.1 | 2 - 3 | 348 | 113 | ||
| GenBank mRNA | BC087741.1 | - | 2 - 3 | 252 | 7314130 | 113 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC100909505
Genomic Map