Assay Details
| Gene Symbol: LOC100909505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750325.3 | XP_003750373.1 | 621 | 72 | ||
| Gene Symbol: Gns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011989.1 | NP_001011989.1 | 4 - 5 | 524 | 72 | |
| XM_006241396.2 | XP_006241458.1 | 4 - 5 | 620 | 72 | ||
| GenBank mRNA | BC087741.1 | - | 4 - 5 | 524 | 7314130 | 72 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100909505
Genomic Map