Assay Details
| Gene Symbol: Tmem173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109122.1 | NP_001102592.1 | 2 - 3 | 359 | 61 | |
| XM_006254601.2 | XP_006254663.1 | 1 - 2 | 326 | 61 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109122.1 transcripts.
Set Membership: |
|

Tmem173
Genomic Map