Assay Details
| Gene Symbol: Emp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007721.1 | NP_001007722.1 | 3 - 4 | 343 | 90 | |
| GenBank mRNA | BC081865.1 | - | 3 - 4 | 343 | 7096470 | 90 |
More Information
Updated Mapping Information
Set Membership: |
|

Emp2
Genomic Map