Assay Details
| Gene Symbol: Emp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007721.1 | NP_001007722.1 | 4 - 5 | 496 | 57 | |
| GenBank mRNA | BC081865.1 | - | 4 - 5 | 496 | 7096470 | 57 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
membrane raft assembly
T cell mediated cytotoxicity
regulation of cell-matrix adhesion
positive regulation of cell-matrix adhesion
regulation of glomerular filtration
actin filament organization
cell adhesion
cell-matrix adhesion
embryo implantation
cell death
cell proliferation
positive regulation of cell proliferation
regulation of endothelial cell migration
cell migration
bleb assembly
activation of protein kinase activity
protein localization to cell surface
blood vessel endothelial cell migration
regulation of kinase activity
early endosome to late endosome transport
regulation of angiogenesis
actin-mediated cell contraction
caveola assembly
protein localization to plasma membrane
positive regulation of integrin-mediated signaling pathway
regulation of vasculogenesis
integrin binding
kinase binding
protein kinase binding
membrane raft assembly
T cell mediated cytotoxicity
regulation of cell-matrix adhesion
positive regulation of cell-matrix adhesion
regulation of glomerular filtration
actin filament organization
cell adhesion
cell-matrix adhesion
embryo implantation
cell death
cell proliferation
positive regulation of cell proliferation
regulation of endothelial cell migration
cell migration
bleb assembly
activation of protein kinase activity
protein localization to cell surface
blood vessel endothelial cell migration
regulation of kinase activity
early endosome to late endosome transport
regulation of angiogenesis
actin-mediated cell contraction
caveola assembly
protein localization to plasma membrane
positive regulation of integrin-mediated signaling pathway
regulation of vasculogenesis
integrin binding
kinase binding
protein kinase binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P011620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717769 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2322 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785723 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334709 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272901 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31506 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P13924 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P13924 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P13924 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P427006 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P13924 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22256 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 33675 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 20821 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10731535 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10731537 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10731532 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10731536 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10731533 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10731534 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10731538 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10731531 | - | - |
| Rat Genome 230 2.0 Array | 1377752_at | - | - |
| Rat Genome 230 2.0 Array | 1373617_at | - | - |
| Rat Genome 230 2.0 Array | 1377752_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1373617_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1377752_at | - | - |
| Affymetrix HT Rat Focus Array | 1373617_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1377752_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1373617_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000000756.rn.1 | TC1000000079.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000000757.rn.1 | TC1000000079.rn.1 | RTA TC |
| Rat Expression Set 230 | 1373617_at | - | - |
| Rat Expression Set 230 | 1373617_PM_at | - | - |
| Rat Expression Set 230 | 1377752_PM_at | - | - |
| Rat Expression Set 230 | 1377752_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10731531 | - | - |
| Rat Genome U34 Set | rc_AI072159_at | - | - |
| Rat Genome U34 Set | rc_AA818807_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20764810 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351017 | - | - |
| RatRef-12 Expression BeadChip | ILMN_53664 | - | - |

Emp2
Genomic Map