Assay Details
| Gene Symbol: Cdip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008360.1 | NP_001008361.1 | 3 - 4 | 238 | 74 | |
| XM_006245811.2 | XP_006245873.1 | 2 - 3 | 249 | 74 | ||
| XM_006245812.2 | XP_006245874.1 | 4 - 5 | 436 | 74 | ||
| XM_006245813.2 | XP_006245875.1 | 3 - 4 | 759 | 74 | ||
| XM_006245814.2 | XP_006245876.1 | 3 - 4 | 362 | 74 | ||
| XM_006245815.2 | XP_006245877.1 | 2 - 3 | 685 | 74 | ||
| XM_006245816.2 | XP_006245878.1 | 2 - 3 | 252 | 74 | ||
| XM_008767493.1 | XP_008765715.1 | 2 - 3 | 1663 | 74 | ||
| XM_008767494.1 | XP_008765716.1 | 2 - 3 | 1914 | 74 | ||
| GenBank mRNA | BC085860.1 | - | 3 - 4 | 238 | 7190340 | 74 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC085860.1, NM_001008360.1, XM_006245811.2, XM_006245812.2, XM_006245813.2, XM_006245814.2, XM_006245815.2, XM_006245816.2, XM_008767493.1, XM_008767494.1 transcripts.
Set Membership: |
|

Cdip1
Genomic Map