Assay Details
| Gene Symbol: Cdip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008360.1 | NP_001008361.1 | 4 - 5 | 348 | 77 | |
| XM_006245811.2 | XP_006245873.1 | 3 - 4 | 359 | 77 | ||
| XM_006245812.2 | XP_006245874.1 | 5 - 6 | 546 | 77 | ||
| XM_006245813.2 | XP_006245875.1 | 4 - 5 | 869 | 77 | ||
| XM_006245814.2 | XP_006245876.1 | 4 - 5 | 472 | 77 | ||
| XM_006245815.2 | XP_006245877.1 | 3 - 4 | 795 | 77 | ||
| XM_006245816.2 | XP_006245878.1 | 3 - 4 | 362 | 77 | ||
| XM_008767493.1 | XP_008765715.1 | 3 - 4 | 1773 | 77 | ||
| XM_008767494.1 | XP_008765716.1 | 3 - 4 | 2024 | 77 | ||
| GenBank mRNA | BC085860.1 | - | 4 - 5 | 348 | 7190340 | 77 |
More Information
Updated Mapping Information
Set Membership: |
|

Cdip1
Genomic Map