Assay Details
| Gene Symbol: Cmpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025655.1 | NP_001020826.1 | 4 - 5 | 637 | 115 | |
| GenBank mRNA | BC098727.1 | - | 4 - 5 | 637 | 7379366 | 115 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cmpk1
Genomic Map