Assay Details
| Gene Symbol: Acot11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271383.1 | NP_001258312.1 | 13 - 14 | 1482 | 59 | |
| XM_008763851.1 | XP_008762073.1 | 13 - 14 | 1502 | 59 | ||
| XM_008763852.1 | XP_008762074.1 | 13 - 14 | 1443 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acot11
Genomic Map