Assay Details
| Gene Symbol: Tuba1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011995.1 | NP_001011995.1 | 1 - 2 | 82 | 55 | |
| GenBank mRNA | BC078829.1 | - | 1 - 2 | 82 | 7112420 | 55 |
| FQ225183.1 | - | 1 - 2 | 103 | 55 | ||
| FQ226218.1 | - | 1 - 2 | 103 | 55 | ||
| FQ235315.1 | - | 1 - 2 | 103 | 55 | ||
| Gene Symbol: LOC100909441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749354.3 | XP_003749402.1 | 1 - 2 | 93 | 55 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC078829.1,NM_001011995.1,XM_003749354.3 transcripts.
This assay is in the 5'UTR region of the BC078829.1,NM_001011995.1,XM_003749354.3 transcripts.
Set Membership: |
|

Tuba1c
Genomic Map