Assay Details
| Gene Symbol: Rnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013222.1 | NP_001013240.1 | 2 - 3 | 301 | 70 | |
| GenBank mRNA | BC089994.1 | - | 2 - 3 | 301 | 7320068 | 70 |
More Information
Updated Mapping Information
Set Membership: |
|

Rnd1
Genomic Map