Assay Details
| Gene Symbol: Pgbd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106198.1 | NP_001099668.1 | 2 - 3 | 706 | 71 | |
| XM_006255807.2 | XP_006255869.1 | 2 - 3 | 793 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pgbd5
Genomic Map