Assay Details
| Gene Symbol: Hsbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173119.2 | NP_775142.1 | 1 - 2 | 136 | 94 | |
| GenBank mRNA | AF522937.1 | - | 1 - 2 | 83 | 94 | |
| BC058131.1 | - | 1 - 2 | 121 | 6919748 | 94 | |
| EX491168.1 | - | 1 - 2 | 136 | 94 | ||
| FQ211891.1 | - | 1 - 2 | 104 | 94 | ||
| FQ212621.1 | - | 1 - 2 | 108 | 94 | ||
| FQ213728.1 | - | 1 - 2 | 78 | 94 | ||
| FQ216175.1 | - | 1 - 2 | 115 | 94 | ||
| FQ229155.1 | - | 1 - 2 | 121 | 94 | ||
| FQ229754.1 | - | 1 - 2 | 95 | 94 | ||
| FQ230047.1 | - | 1 - 2 | 93 | 94 | ||
| FQ230722.1 | - | 1 - 2 | 95 | 94 | ||
| FQ233773.1 | - | 1 - 2 | 94 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsbp1
Genomic Map