Assay Details
| Gene Symbol: Rfx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105944.1 | NP_001099414.1 | 8 - 9 | 1124 | 56 | |
| XM_006255229.2 | XP_006255291.1 | 8 - 9 | 1276 | 56 | ||
| XM_006255231.2 | XP_006255293.1 | 8 - 9 | 1276 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rfx1
Genomic Map