Assay Details
| Gene Symbol: Klf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107164.1 | NP_001100634.1 | 1 - 2 | 192 | 56 | |
| XM_006255254.2 | XP_006255316.1 | 1 - 2 | 280 | 56 | ||
| GenBank mRNA | BC158887.1 | - | 1 - 2 | 122 | 56 | |
| FQ232766.1 | - | 1 - 2 | 152 | 56 | ||
| FQ235088.1 | - | 1 - 2 | 154 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Klf1
Genomic Map