Assay Details
| Gene Symbol: Chst5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006255633.2 | XP_006255695.1 | 2 - 3 | 921 | 108 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006255633.2 transcripts.
Set Membership: |
|

Chst5
Genomic Map