Assay Details
| Gene Symbol: Mri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010947.1 | NP_001010947.1 | 3 - 4 | 766 | 75 | |
| GenBank mRNA | BC089060.1 | - | 3 - 4 | 766 | 7320396 | 75 |
| FQ213644.1 | - | 3 - 4 | 758 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mri1
Genomic Map