Assay Details
| Gene Symbol: Bola1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001071776.1 | NP_001065244.1 | 1 - 2 | 64 | 75 | |
| GenBank mRNA | DQ912022.1 | - | 1 - 2 | 64 | 75 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the DQ912022.1,NM_001071776.1 transcripts.
Set Membership: |
|

Bola1
Genomic Map