Assay Details
| Gene Symbol: Scara3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108870.1 | NP_001102340.1 | 2 - 3 | 355 | 78 | |
| GenBank mRNA | FQ211593.1 | - | 2 - 3 | 372 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scara3
Genomic Map