Assay Details
| Gene Symbol: Scara3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108870.1 | NP_001102340.1 | 5 - 6 | 1610 | 79 | |
| GenBank mRNA | FQ211593.1 | - | 5 - 6 | 1412 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P139549 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P139544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779520 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272398 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373313 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98859 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P139544 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P139549 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P139544 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P139549 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P139549 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P139544 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P305272 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23215 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10784580 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784583 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784581 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784584 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784582 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10784579 | - | - |
| Rat Genome 230 2.0 Array | 1382081_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382081_at | - | - |
| Affymetrix HT Rat Focus Array | 1382081_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1382081_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1500012401.rn.1 | TC1500001852.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500012399.rn.1 | TC1500001852.rn.1 | RTA TC |
| Rat Expression Set 230 | 1382081_PM_at | - | - |
| Rat Expression Set 230 | 1382081_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784579 | - | - |
| Rat Genome U34 Set | rc_AA924758_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21259313 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365032 | - | - |

Scara3
Genomic Map