Assay Details
| Gene Symbol: Spryd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009635.1 | NP_001009635.1 | 1 - 2 | 199 | 76 | |
| GenBank mRNA | BC088471.1 | - | 1 - 2 | 199 | 7128422 | 76 |
More Information
Updated Mapping Information
Set Membership: |
|

Spryd7
Genomic Map