Assay Details
| Gene Symbol: Zfp786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127594.1 | NP_001121066.1 | 2 - 3 | 185 | 111 | |
| XM_006236413.2 | XP_006236475.1 | 2 - 3 | 242 | 111 | ||
| XM_006236414.2 | XP_006236476.1 | 2 - 3 | 328 | 111 | ||
| GenBank mRNA | BC166479.1 | - | 2 - 3 | 185 | 9102251 | 111 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the XM_006236413.2,XM_006236414.2 transcripts.
Set Membership: |
|

Zfp786
Genomic Map