Assay Details
| Gene Symbol: Fam188b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_231787.7 | XP_231787.5 | 17 - 18 | 2361 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam188b| Gene Symbol: Fam188b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_231787.7 | XP_231787.5 | 17 - 18 | 2361 | 65 | |
Set Membership: |
|