Assay Details
| Gene Symbol: Fam188b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_231787.7 | XP_231787.5 | 1 - 2 | 200 | 105 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_231787.7 transcripts.
Set Membership: |
|

Fam188b
Genomic Map