Assay Details
| Gene Symbol: Rnf168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127597.2 | NP_001121069.2 | 4 - 5 | 702 | 98 | |
| GenBank mRNA | BC166869.1 | - | 4 - 5 | 702 | 9030307 | 98 |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf168
Genomic Map