Assay Details
| Gene Symbol: Gcsam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001064866.4 | XP_001064866.2 | 3 - 4 | 351 | 75 | |
| XM_006248351.2 | XP_006248413.1 | 2 - 3 | 247 | 75 | ||
More Information
Set Membership: |
|

Gcsam
Genomic Map