Assay Details
| Gene Symbol: Gcsam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001064866.4 | XP_001064866.2 | 5 - 6 | 434 | 105 | |
| XM_006248351.2 | XP_006248413.1 | 4 - 5 | 330 | 105 | ||
More Information
Set Membership: |
|

Gcsam
Genomic Map