Assay Details
| Gene Symbol: Rap2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106950.2 | NP_001100420.1 | 1 - 2 | 386 | 141 | |
| GenBank mRNA | BP503228.1 | - | 1 - 2 | 400 | 141 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rap2c
Genomic Map