Assay Details
| Gene Symbol: Zfp280c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006257541.2 | XP_006257603.1 | 2392 | 136 | ||
| XM_006257542.2 | XP_006257604.1 | 2291 | 136 | |||
| XM_008773539.1 | XP_008771761.1 | 18 - 19 | 2279 | 136 | ||
| XM_008773540.1 | XP_008771762.1 | 2248 | 136 | |||
| XM_008773541.1 | XP_008771763.1 | 17 - 18 | 2243 | 136 | ||
| XM_008773542.1 | XP_008771764.1 | 16 - 17 | 3273 | 136 | ||
More Information
Set Membership: |
|

Zfp280c
Genomic Map