Assay Details
| Gene Symbol: Zfp280c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006257541.2 | XP_006257603.1 | 599 | 83 | ||
| XM_006257542.2 | XP_006257604.1 | 498 | 83 | |||
| XM_008773539.1 | XP_008771761.1 | 6 - 7 | 486 | 83 | ||
| XM_008773540.1 | XP_008771762.1 | 455 | 83 | |||
| XM_008773541.1 | XP_008771763.1 | 5 - 6 | 450 | 83 | ||
| XM_008773542.1 | XP_008771764.1 | 4 - 5 | 1480 | 83 | ||
More Information
Set Membership: |
|

Zfp280c
Genomic Map