Assay Details
| Gene Symbol: Zfp280c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006257541.2 | XP_006257603.1 | 1209 | 117 | ||
| XM_006257542.2 | XP_006257604.1 | 1108 | 117 | |||
| XM_008773539.1 | XP_008771761.1 | 10 - 11 | 1096 | 117 | ||
| XM_008773540.1 | XP_008771762.1 | 1065 | 117 | |||
| XM_008773541.1 | XP_008771763.1 | 9 - 10 | 1060 | 117 | ||
| XM_008773542.1 | XP_008771764.1 | 8 - 9 | 2090 | 117 | ||
More Information
Set Membership: |
|

Zfp280c
Genomic Map