Assay Details
| Gene Symbol: Itgb3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013213.1 | NP_001013231.1 | 6 - 7 | 752 | 126 | |
| XM_008763882.1 | XP_008762104.1 | 4 - 5 | 569 | 126 | ||
| XM_008763883.1 | XP_008762105.1 | 7 - 8 | 494 | 126 | ||
| XM_008763884.1 | XP_008762106.1 | 6 - 7 | 448 | 126 | ||
| GenBank mRNA | BC086363.1 | - | 6 - 7 | 752 | 7304501 | 126 |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P036237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678351 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78534 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776093 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2650 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P036237 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P036237 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P036237 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P274641 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46791 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 37029 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 2447 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 62280 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10878273 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878277 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878274 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878279 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878280 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878276 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878278 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878275 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10878272 | - | - |
| Rat Genome 230 2.0 Array | 1390604_PM_s_at | - | - |
| Rat Genome 230 2.0 Array | 1377182_at | - | - |
| Rat Genome 230 2.0 Array | 1377182_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1390604_s_at | - | - |
| Affymetrix HT Rat Focus Array | 1390604_s_at | - | - |
| Affymetrix HT Rat Focus Array | 1377182_at | - | - |
| Affymetrix HT Rat Focus Array | 1390604_PM_s_at | - | - |
| Affymetrix HT Rat Focus Array | 1377182_PM_at | - | - |
| Rat Expression Set 230 | 1377182_at | - | - |
| Rat Expression Set 230 | 1390604_s_at | - | - |
| Rat Expression Set 230 | 1390604_PM_s_at | - | - |
| Rat Expression Set 230 | 1377182_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878272 | - | - |
| Rat Genome U34 Set | rc_AA818179_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373985 | - | - |

Itgb3bp
Genomic Map