Assay Details
| Gene Symbol: Erf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170335.1 | NP_001163806.1 | 1 - 2 | 186 | 64 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001170335.1 transcripts.
Set Membership: |
|

Erf
Genomic Map