Assay Details
| Gene Symbol: Rinl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106241.1 | NP_001099711.1 | 4 - 5 | 421 | 82 | |
| XM_006228653.2 | XP_006228715.1 | 4 - 5 | 673 | 82 | ||
| XM_006228654.2 | XP_006228716.1 | 4 - 5 | 661 | 82 | ||
| XM_006228655.2 | XP_006228717.1 | 4 - 5 | 673 | 82 | ||
| XM_006228656.2 | XP_006228718.1 | 4 - 5 | 673 | 82 | ||
| XM_006228657.2 | XP_006228719.1 | 3 - 4 | 396 | 82 | ||
| XM_008759120.1 | XP_008757342.1 | 3 - 4 | 538 | 82 | ||
| XM_008759121.1 | XP_008757343.1 | 4 - 5 | 506 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rinl
Genomic Map