Assay Details
| Gene Symbol: Wtip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759260.1 | XP_008757482.1 | 4 - 5 | 335 | 73 | |
| GenBank mRNA | BC167052.1 | - | 4 - 5 | 350 | 73 | |
More Information
Set Membership: |
|

Wtip
Genomic Map