Assay Details
| Gene Symbol: Wtip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759260.1 | XP_008757482.1 | 5 - 6 | 460 | 77 | |
| GenBank mRNA | BC167052.1 | - | 5 - 6 | 475 | 77 | |
More Information
Set Membership: |
|

Wtip
Genomic Map