Assay Details
| Gene Symbol: Ano5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006229266.2 | XP_006229328.1 | 18 - 19 | 3336 | 63 | |
| XM_006229268.2 | XP_006229330.1 | 18 - 19 | 3227 | 63 | ||
More Information
Set Membership: |
|

Ano5
Genomic Map