Assay Details
| Gene Symbol: March1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135838.1 | NP_001129310.1 | 3 - 4 | 229 | 65 | |
| XM_008771132.1 | XP_008769354.1 | 1 - 2 | 541 | 65 | ||
| XM_008771133.1 | XP_008769355.1 | 1 - 2 | 367 | 65 | ||
| XM_008771134.1 | XP_008769356.1 | 1 - 2 | 448 | 65 | ||
| GenBank mRNA | FQ230739.1 | - | 1 - 2 | 310 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

March1
Genomic Map