Assay Details
| Gene Symbol: Ccl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008513.1 | NP_001008513.1 | 2 - 3 | 237 | 87 | |
| GenBank mRNA | BC086571.1 | - | 2 - 3 | 237 | 7304213 | 87 |
| FQ218040.1 | - | 2 - 3 | 261 | 87 | ||
| FQ220091.1 | - | 2 - 3 | 261 | 87 | ||
| FQ223815.1 | - | 2 - 3 | 263 | 87 | ||
| FQ225478.1 | - | 2 - 3 | 261 | 87 | ||
| FQ226079.1 | - | 2 - 3 | 259 | 87 | ||
| FQ233622.1 | - | 2 - 3 | 261 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccl21
Genomic Map